missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5332503

 View more versions of this product

Catalog No. AAH5332503



Specifications

Specifications

4-Chloro-3-fluorobenzeneboronic acid
1g
C6H5BClFO2
4-chloro-3-fluorophenyl boronic acid, 4-chloro-3-fluorobenzeneboronic acid, 3-fluoro-4-chlorophenylboronic acid, 4-chloro-3-fluoro-phenyl boronic acid, boronic acid, 4-chloro-3-fluorophenyl, 4-chloro-3-fluorobenzeneboronicacid, pubchem1776, ksc174k0r, acmc-1c195, 4-chloro3-fluorophenylboronic acid
B(C1=CC(=C(C=C1)Cl)F)(O)O
174.362
174.37
198°C to 204°C
137504-86-0
MFCD01319010
CMJQIHGBUKZEHP-UHFFFAOYSA-N
(4-chloro-3-fluorophenyl)boronic acid
2778656
97%

Chemical Identifiers

137504-86-0
174.362
CMJQIHGBUKZEHP-UHFFFAOYSA-N
2778656
B(C1=CC(=C(C=C1)Cl)F)(O)O
C6H5BClFO2
MFCD01319010
4-chloro-3-fluorophenyl boronic acid, 4-chloro-3-fluorobenzeneboronic acid, 3-fluoro-4-chlorophenylboronic acid, 4-chloro-3-fluoro-phenyl boronic acid, boronic acid, 4-chloro-3-fluorophenyl, 4-chloro-3-fluorobenzeneboronicacid, pubchem1776, ksc174k0r, acmc-1c195, 4-chloro3-fluorophenylboronic acid
(4-chloro-3-fluorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents