Please login to your online account to display your discounted pricing

4-Chloro-3-fluorobenzaldehyde, 98%, ACROS Organics™

$125.57 - $125.57

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-fluorobenzaldehyde
CAS 5527-95-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4ClFO
Linear Formula ClC6H3(F)CHO
Formula Weight 158.56
MDL Number MFCD00143288
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC294140250

  SDS
thermo fisher scientific
AC294140250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC294140050

  SDS
thermo fisher scientific
294140050
Glass bottle 5g Each for $125.57
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 158.56
Physical Form Crystalline Solid
Quantity 25g
Color White to yellow
CAS Max % 100.0
Chemical Name or Material 4-Chloro-3-fluorobenzaldehyde
Assay Percent Range 98%
CAS 5527-95-7
MDL Number MFCD00143288
CAS Min % 97.5
Packaging Glass bottle
Linear Formula ClC6H3(F)CHO
Infrared Spectrum Authentic
Molecular Formula C7H4ClFO
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.