missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-fluorobenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 294140050

 View more versions of this product

Catalog No. AC294140050



Description

Description

Specifications

Specifications

4-Chloro-3-fluorobenzaldehyde
Glass bottle
5g
97.5
98%
ClC6H3(F)CHO
3-fluoro-4-chlorobenzaldehyde, 4-chloro-3-fluoro benzaldehyde, 4-chloro-3-fluoro-benzaldehyde, 4-chloro-3-fluorobenz-aldehyde, acmc-1amt7, azmdwrptdcifrd-uhfffaoysa, benzaldehyde, 4-chloro-3-fluoro, ksc494a8n, pubchem1412
C1=CC(=C(C=C1C=O)F)Cl
158.556
158.56
≥97.5%
Authentic
White to Yellow
5527-95-7
100.0
C7H4ClFO
MFCD00143288
AZMDWRPTDCIFRD-UHFFFAOYSA-N
4-chloro-3-fluorobenzaldehyde
2724985
Crystalline Solid

Chemical Identifiers

5527-95-7
158.556
AZMDWRPTDCIFRD-UHFFFAOYSA-N
2724985
C1=CC(=C(C=C1C=O)F)Cl
C7H4ClFO
MFCD00143288
3-fluoro-4-chlorobenzaldehyde, 4-chloro-3-fluoro benzaldehyde, 4-chloro-3-fluoro-benzaldehyde, 4-chloro-3-fluorobenz-aldehyde, acmc-1amt7, azmdwrptdcifrd-uhfffaoysa, benzaldehyde, 4-chloro-3-fluoro, ksc494a8n, pubchem1412
4-chloro-3-fluorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents