Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-fluorobenzaldehyde, 98%

$37.68 - $37.68

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-fluorobenzaldehyde
CAS 5527-95-7
Formula Weight 158.56
MDL Number MFCD00143288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4136503

  SDS
alfa aesar™
4136503
1g Each for $37.68

AA4136514

  SDS
alfa aesar™
4136514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4136506

  SDS
alfa aesar™
4136506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 4-Chloro-3-fluorobenzaldehyde
Flash Point 96° (206°F)
Sensitivity Air Sensitive
Color White
Melting Point 46° to 48°C
Quantity 1g
CAS 5527-95-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
Assay 98%
Formula Weight 158.56
MDL Number MFCD00143288
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.