Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3,5-dinitrobenzotrifluoride, 97%

$61.78 - $61.78

Chemical Identifiers

Chemical Name or Material 4-Chloro-3,5-dinitrobenzotrifluoride
CAS 393-75-9
Molecular Formula C7H2ClF3N2O4
Formula Weight 270.55
MDL Number MFCD00007068
UN Number UN2811
Beilstein 1220937
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1044606

  SDS
alfa aesar™
A1044606
5g Each for $61.78
Description & Specifications

Specifications

Formula Weight 270.55
Health Hazard 3 GHS P Statement
P261-P305+P351+P338-P361-P302+P350-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 393-75-9
Quantity 5g
Assay 97%
Chemical Name or Material 4-Chloro-3,5-dinitrobenzotrifluoride
Boiling Point >250°C
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number XS9065000
Melting Point 53° to 58°C
Health Hazard 1 Danger
Flash Point 126°C (258°F)
MDL Number MFCD00007068
TSCA TSCA
Molecular Formula C7H2ClF3N2O4
Beilstein 1220937
Health Hazard 2 GHS H Statement
H310-H302-H315-H319-H335
Fatal in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
4-Chloro-3,5-dinitrobenzotrifluoride is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents, strong bases. Stable at room temperature.

Safety and Handling

Danger

GHS P Statement
P261-P305+P351+P338-P361-P302+P350-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H310-H302-H315-H319-H335
Fatal in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.