Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3,5-dinitrobenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3,5-dinitrobenzonitrile
CAS 1930-72-9
Molecular Formula C7H2ClN3O4
Formula Weight 227.56
MDL Number MFCD00007067
UN Number UN3439
Beilstein 1990451
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0341303

  SDS
alfa aesar™
L0341303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0341306

  SDS
alfa aesar™
L0341306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.56
CAS 1930-72-9
Chemical Name or Material 4-Chloro-3,5-dinitrobenzonitrile
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Beilstein 1990451
MDL Number MFCD00007067
Quantity 1g
Molecular Formula C7H2ClN3O4
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
UN Number UN3439
Melting Point 140° to 142°C
RTECS Number DI2990000

Applications
4-Chloro-3,5-dinitrobenzonitrile is may be used in the synthesis of 3-nitro-4-thiocyanobenzonitryl. It is also used as Pharmaceutical intermediates .

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong bases.Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.