missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3,5-dinitrobenzoic acid, 99%

Manufacturer:  Alfa Aesar™ A1237322

 View more versions of this product

Catalog No. AAA1237322



Specifications

Specifications

4-Chloro-3,5-dinitrobenzoic acid
315°C
100g
99%
MFCD00007080
PCTFIHOVQYYAMH-UHFFFAOYSA-N
246.559
246.56
186°C (367°F)
159°C to 162°C
118-97-8
C7H3ClN2O6
668253
C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(=O)O
8377

Chemical Identifiers

118-97-8
246.559
PCTFIHOVQYYAMH-UHFFFAOYSA-N
C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(=O)O
C7H3ClN2O6
MFCD00007080
8377
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents