missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3,5-dimethylphenol, 99%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC109060025

 View more versions of this product

Catalog No. AC109060025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Chloro-3, 5-dimethylphenol
88-04-0
100.0
113.0°C to 116.0°C
132°C
Authentic
ClC6H2(CH3)2OH
06, II, 463
15, 2182
2.5kg
OSDLLIBGSJNGJE-UHFFFAOYSA-N
4-chloro-3,5-dimethylphenol
2723
156.61
99%
99%
98.5
White to Beige
246.0°C
98.5% min. (GC)
C8H9ClO
MFCD00002324
Plastic bottle
2-chloro-m-xylenol, 4-chloro-3,5-xylenol, 4-chloro-m-xylenol, benzytol, chloroxylenol, desson, dettol, ottasept, p-chloro-m-xylenol, pcmx
Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alkali hydroxide solutions,soluble in ethanol,ether and benzene
CC1=CC(=CC(=C1Cl)C)O
156.61
CHEBI:34393
Crystalline Powder or Crystals

Chemical Identifiers

88-04-0
156.61
OSDLLIBGSJNGJE-UHFFFAOYSA-N
2723
4-chloro-3,5-dimethylphenol
C8H9ClO
MFCD00002324
2-chloro-m-xylenol, 4-chloro-3,5-xylenol, 4-chloro-m-xylenol, benzytol, chloroxylenol, desson, dettol, ottasept, p-chloro-m-xylenol, pcmx
CHEBI:34393
CC1=CC(=CC(=C1Cl)C)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse

GHS Signal Word: Warning

EINECSNumber : 201-793-8

RTECSNumber : ZE6850000

TSCA : TSCA

SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only