Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3,5-difluoropyridine, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3,5-difluoropyridine
CAS 851178-97-7
Molecular Formula C5H2ClF2N
Formula Weight 149.53g/mol
MDL Number MFCD09998885
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3161103

 
alfa aesar™
H31611-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH31611MD

 
alfa aesar™
H31611-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 124°C (760mmHg)
Quantity 1g
Flash Point 28.8°C(84°F)
Chemical Name or Material 4-Chloro-3,5-difluoropyridine
CAS 851178-97-7
Assay 97%
Molecular Formula C5H2ClF2N
Formula Weight 149.53g/mol
MDL Number MFCD09998885
UN Number UN1993
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.