missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-3-(4-morpholinylcarbonyl)benzeneboronic acid, 98%, Thermo Scientific™
$205.82 - $205.82
Chemical Identifiers
CAS | 871332-71-7 |
---|---|
Molecular Formula | C11H13BClNO4 |
Molecular Weight (g/mol) | 269.488 |
MDL Number | MFCD07783863 |
InChI Key | SWIHOHJBUZBINE-UHFFFAOYSA-N |
Synonym | 4-chloro-3-morpholine-4-carbonyl phenyl boronic acid, 4-chloro-3-n-morpholinecarbonyl phenylboronic acid, 4-chloro-3-morpholine-4-carbonyl phenylboronic acid, acmc-209qh1, 4-chloro-3-4-morpholinylcarbonyl benzeneboronic acid, 4-chloro-3-4-morpholinylcarbonyl phenylboronic acid, boronic acid,b-4-chloro-3-4-morpholinylcarbonyl phenyl |
PubChem CID | 44886934 |
IUPAC Name | [4-chloro-3-(morpholine-4-carbonyl)phenyl]boronic acid |
SMILES | B(C1=CC(=C(C=C1)Cl)C(=O)N2CCOCC2)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH53336MD
|
Thermo Scientific Chemicals
H53336MD |
250 mg |
Each for $205.82
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Chloro-3-(4-morpholinylcarbonyl)benzeneboronic ac is involved in suzuki-Miyaura coupling reaction.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
871332-71-7 | |
269.488 | |
SWIHOHJBUZBINE-UHFFFAOYSA-N | |
44886934 | |
B(C1=CC(=C(C=C1)Cl)C(=O)N2CCOCC2)(O)O |
C11H13BClNO4 | |
MFCD07783863 | |
4-chloro-3-morpholine-4-carbonyl phenyl boronic acid, 4-chloro-3-n-morpholinecarbonyl phenylboronic acid, 4-chloro-3-morpholine-4-carbonyl phenylboronic acid, acmc-209qh1, 4-chloro-3-4-morpholinylcarbonyl benzeneboronic acid, 4-chloro-3-4-morpholinylcarbonyl phenylboronic acid, boronic acid,b-4-chloro-3-4-morpholinylcarbonyl phenyl | |
[4-chloro-3-(morpholine-4-carbonyl)phenyl]boronic acid |
Specifications
871332-71-7 | |
C11H13BClNO4 | |
250 mg | |
Slightly soluble in water. | |
B(C1=CC(=C(C=C1)Cl)C(=O)N2CCOCC2)(O)O | |
269.488 | |
253.49 | |
4-Chloro-3-(4-morpholinylcarbonyl)benzeneboronic acid |
146°C to 150°C | |
MFCD07783863 | |
4-chloro-3-morpholine-4-carbonyl phenyl boronic acid, 4-chloro-3-n-morpholinecarbonyl phenylboronic acid, 4-chloro-3-morpholine-4-carbonyl phenylboronic acid, acmc-209qh1, 4-chloro-3-4-morpholinylcarbonyl benzeneboronic acid, 4-chloro-3-4-morpholinylcarbonyl phenylboronic acid, boronic acid,b-4-chloro-3-4-morpholinylcarbonyl phenyl | |
SWIHOHJBUZBINE-UHFFFAOYSA-N | |
[4-chloro-3-(morpholine-4-carbonyl)phenyl]boronic acid | |
44886934 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures