missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(2-chlorophenylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5490103

 View more versions of this product

Catalog No. AAH5490103



Specifications

Specifications

4-Chloro-3-(2-chlorophenylcarbamoyl)benzeneboronic acid
1449132-45-9
MFCD20265367
NDPXQPNFRRTCMV-UHFFFAOYSA-N
[4-chloro-3-[(2-chlorophenyl)carbamoyl]phenyl]boronic acid
73996020
97%
1g
C13H10BCl2NO3
4-chloro-3-2-chlorophenyl carbamoyl phenylboronic acid
B(C1=CC(=C(C=C1)Cl)C(=O)NC2=CC=CC=C2Cl)(O)O
309.937
309.94

Chemical Identifiers

1449132-45-9
309.937
NDPXQPNFRRTCMV-UHFFFAOYSA-N
73996020
B(C1=CC(=C(C=C1)Cl)C(=O)NC2=CC=CC=C2Cl)(O)O
C13H10BCl2NO3
MFCD20265367
4-chloro-3-2-chlorophenyl carbamoyl phenylboronic acid
[4-chloro-3-[(2-chlorophenyl)carbamoyl]phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents