Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)quinoline, 98%

$172.41 - $172.41

Chemical Identifiers

Chemical Name or Material 4-Chloro-2- (trifluoromethyl)quinoline
CAS 1701-24-2
Molecular Formula C10H5ClF3N
Formula Weight 231.61
MDL Number MFCD00153105
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1083406

  SDS
alfa aesar™
A1083406
5g Each for $172.41
Description & Specifications

Specifications

Formula Weight 231.61
Chemical Name or Material 4-Chloro-2- (trifluoromethyl)quinoline
Assay 98%
Molecular Formula C10H5ClF3N
MDL Number MFCD00153105
CAS 1701-24-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 107°C (225°F)
Melting Point 31° to 33°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.