Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)quinazoline, 98%

$129.92 - $449.47

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-(trifluoromethyl)quinazoline
CAS 52353-35-2
Molecular Formula C9H4ClF3N2
Formula Weight 232.59
MDL Number MFCD00793683
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5052903

  SDS
alfa aesar™
H5052903
1g Each for $449.47

AAH50529MD

  SDS
alfa aesar™
H50529MD
250mg Each for $129.92
Description & Specifications

Specifications

Formula Weight 232.59
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)quinazoline
Assay 98%
Molecular Formula C9H4ClF3N2
MDL Number MFCD00793683
CAS 52353-35-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g

Applications
4-Chloro-2-(trifluoromethyl)quinazoline is used as pharmaceutical intermediates. It exhibits antiplasmodial properties.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.