Learn More
4-Chloro-2-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
$175.51 - $698.82
Chemical Identifiers
CAS | 131748-14-6 |
---|---|
Molecular Formula | C6H3ClF3N |
Molecular Weight (g/mol) | 181.54 |
MDL Number | MFCD07368047 |
InChI Key | FZINIBTZNSPWQR-UHFFFAOYSA-N |
Synonym | 4-chloro-2-trifluoromethyl pyridine, 2-trifluoromethyl-4-chloropyridine, pyridine,4-chloro-2-trifluoromethyl, pubchem15520, acmc-209bnu, 4-chloro-2-trifluoromethyl-pyridine, 2-trifluoro methyl-4-chloro pyridine |
PubChem CID | 14761442 |
IUPAC Name | 4-chloro-2-(trifluoromethyl)pyridine |
SMILES | FC(F)(F)C1=NC=CC(Cl)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6483303
|
Thermo Scientific Chemicals
H6483303 |
1 g |
Each for $175.51
|
|
|||||
AAH6483306
|
Thermo Scientific Chemicals
H6483306 |
5 g |
Each for $698.82
|
|
|||||
Description
Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Chemical Identifiers
131748-14-6 | |
181.54 | |
FZINIBTZNSPWQR-UHFFFAOYSA-N | |
14761442 | |
FC(F)(F)C1=NC=CC(Cl)=C1 |
C6H3ClF3N | |
MFCD07368047 | |
4-chloro-2-trifluoromethyl pyridine, 2-trifluoromethyl-4-chloropyridine, pyridine,4-chloro-2-trifluoromethyl, pubchem15520, acmc-209bnu, 4-chloro-2-trifluoromethyl-pyridine, 2-trifluoro methyl-4-chloro pyridine | |
4-chloro-2-(trifluoromethyl)pyridine |
Specifications
131748-14-6 | |
C6H3ClF3N | |
1 g | |
4-chloro-2-trifluoromethyl pyridine, 2-trifluoromethyl-4-chloropyridine, pyridine,4-chloro-2-trifluoromethyl, pubchem15520, acmc-209bnu, 4-chloro-2-trifluoromethyl-pyridine, 2-trifluoro methyl-4-chloro pyridine | |
FZINIBTZNSPWQR-UHFFFAOYSA-N | |
4-chloro-2-(trifluoromethyl)pyridine | |
14761442 | |
98% | |
4-Chloro-2-(trifluoromethyl)pyridine |
Colorless | |
MFCD07368047 | |
UN2810 | |
Slightly soluble in water. | |
FC(F)(F)C1=NC=CC(Cl)=C1 | |
181.54 | |
181.54 | |
Liquid |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only