missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-(trifluoromethyl)phenylboronic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC445380010

 View more versions of this product

Catalog No. AC445380010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Chloro-2-(trifluoromethyl)phenylboronic acid
1g
94% min. (HPLC)
2-chloro-4-trifluoromethyl-phenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethyl phenylboronicacid, 4-chloro-2-trifluoromethylphenylboronic acid, acmc-20aic7, boronic acid, 4-chloro-2-trifluoromethyl phenyl, pubchem5158
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
224.38
224.38
Glass Bottle
313545-41-4
C7H5BClF3O2
YAPBOBGBYQQYHX-UHFFFAOYSA-N
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid
2763244
95%

Chemical Identifiers

313545-41-4
224.38
2-chloro-4-trifluoromethyl-phenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethyl phenylboronicacid, 4-chloro-2-trifluoromethylphenylboronic acid, acmc-20aic7, boronic acid, 4-chloro-2-trifluoromethyl phenyl, pubchem5158
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid
C7H5BClF3O2
YAPBOBGBYQQYHX-UHFFFAOYSA-N
2763244
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

SDS Documents