missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-(trifluoromethyl)phenylboronic acid, 95%, ACROS Organics™

Chemical Identifiers

CAS 313545-41-4
Molecular Formula C7H5BClF3O2
Molecular Weight (g/mol) 224.38
InChI Key YAPBOBGBYQQYHX-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl-phenylboronic acid
PubChem CID 2763244
IUPAC Name [4-chloro-2-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC445380010 SDS View Documents Thermo Fisher Scientific
AC445380010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC445380050 SDS View Documents Thermo Fisher Scientific
AC445380050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

313545-41-4
224.38
2-chloro-4-trifluoromethyl-phenylboronic acid
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid
C7H5BClF3O2
YAPBOBGBYQQYHX-UHFFFAOYSA-N
2763244
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O

Specifications

Glass Bottle
313545-41-4
C7H5BClF3O2
YAPBOBGBYQQYHX-UHFFFAOYSA-N
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid
2763244
95%
1g
94% min. (HPLC)
2-chloro-4-trifluoromethyl-phenylboronic acid
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O
224.38
224.38
4-Chloro-2-(trifluoromethyl)phenylboronic acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning