missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-2-(trifluoromethyl)phenylboronic acid, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC445380050
Specifications
4-Chloro-2-(trifluoromethyl)phenylboronic acid | |
313545-41-4 | |
C7H5BClF3O2 | |
YAPBOBGBYQQYHX-UHFFFAOYSA-N | |
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid | |
2763244 | |
95% |
5g | |
94% min. (HPLC) | |
2-chloro-4-trifluoromethyl-phenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethyl phenylboronicacid, 4-chloro-2-trifluoromethylphenylboronic acid, acmc-20aic7, boronic acid, 4-chloro-2-trifluoromethyl phenyl, pubchem5158 | |
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O | |
224.38 | |
224.38 |
Chemical Identifiers
313545-41-4 | |
224.38 | |
2-chloro-4-trifluoromethyl-phenylboronic acid, 4-chloro-2-trifluoromethyl benzeneboronic acid, 4-chloro-2-trifluoromethyl phenyl boronic acid, 4-chloro-2-trifluoromethyl phenylboronic acid, 4-chloro-2-trifluoromethyl phenylboronicacid, 4-chloro-2-trifluoromethylphenylboronic acid, acmc-20aic7, boronic acid, 4-chloro-2-trifluoromethyl phenyl, pubchem5158 | |
[4-chloro-2-(trifluoromethyl)phenyl]boronic acid |
C7H5BClF3O2 | |
YAPBOBGBYQQYHX-UHFFFAOYSA-N | |
2763244 | |
B(C1=C(C=C(C=C1)Cl)C(F)(F)F)(O)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning