Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)benzoic acid, 97%

$91.46 - $506.27

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzoic acid
CAS 142994-09-0
Molecular Formula C8H4ClF3O2
Formula Weight 224.57
MDL Number MFCD03094162
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2643103

  SDS
alfa aesar™
H2643103
1g Each for $91.46

AAH2643109

  SDS
alfa aesar™
H2643109
10g Each for $506.27
Description & Specifications

Specifications

Formula Weight 224.57
Molecular Formula C8H4ClF3O2
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzoic acid
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
MDL Number MFCD03094162
CAS 142994-09-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.