Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-(trifluoromethyl)benzaldehyde, 97%

$91.46 - $525.86

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzaldehyde
CAS 320-43-4
Molecular Formula C8H4ClF3O
Formula Weight 208.57
MDL Number MFCD03092987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2636009

  SDS
alfa aesar™
H2636009
10g Each for $525.86

AAH2636003

  SDS
alfa aesar™
H2636003
1g Each for $91.46
Description & Specifications

Specifications

Formula Weight 208.57
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Quantity 10g
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzaldehyde
CAS 320-43-4
Molecular Formula C8H4ClF3O
MDL Number MFCD03092987
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.