missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-(trifluoromethoxy)benzoic acid, 97%

Chemical Identifiers

CAS 1261605-70-2
Molecular Formula C8H4ClF3O3
Molecular Weight (g/mol) 240.562
MDL Number MFCD18393801
InChI Key SSORZLDIKSZDTH-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethoxy benzoic acid
PubChem CID 66523556
IUPAC Name 4-chloro-2-(trifluoromethoxy)benzoic acid
SMILES C1=CC(=C(C=C1Cl)OC(F)(F)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3340303 View Documents Alfa Aesar™
H3340303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH3340306 View Documents Alfa Aesar™
H3340306
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1261605-70-2
240.562
SSORZLDIKSZDTH-UHFFFAOYSA-N
66523556
C1=CC(=C(C=C1Cl)OC(F)(F)F)C(=O)O
C8H4ClF3O3
MFCD18393801
4-chloro-2-trifluoromethoxy benzoic acid
4-chloro-2-(trifluoromethoxy)benzoic acid

Specifications

1g
C8H4ClF3O3
4-chloro-2-trifluoromethoxy benzoic acid
C1=CC(=C(C=C1Cl)OC(F)(F)F)C(=O)O
240.562
240.56
4-Chloro-2-(trifluoromethoxy)benzoic acid
1261605-70-2
MFCD18393801
SSORZLDIKSZDTH-UHFFFAOYSA-N
4-chloro-2-(trifluoromethoxy)benzoic acid
66523556
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures