missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-(trifluoromethoxy)aniline, TCI America™

Manufacturer:  TCI America C33741G

 View more versions of this product

Catalog No. C33741G



Specifications

Specifications

4-Chloro-2-(trifluoromethoxy)aniline
White-Yellow
1g
C7H5ClF3NO
4-chloro-2-trifluoromethoxy aniline, 4-chloro-2-trifluoromethoxy phenylamine, benzenamine, 4-chloro-2-trifluoromethoxy, pubchem2960, chlorotrifluoromethoxyphenylamine, benzenamine,4-chloro-2-trifluoromethoxy, 2-amino-5-chlorophenyl trifluoromethyl ether, 2-amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-2-trifluoromethoxy aniline; 4-chloro-2-trifluoromethoxy phenylamine
C1=CC(=C(C=C1Cl)OC(F)(F)F)N
211.568
211.57
≥98.0% (GC)
82°C
33°C
175205-77-3
MFCD00153102
UCFTYLMKCJPWBX-UHFFFAOYSA-N
4-chloro-2-(trifluoromethoxy)aniline
2781526
Crystal-Lump at 20°C

Chemical Identifiers

175205-77-3
211.568
UCFTYLMKCJPWBX-UHFFFAOYSA-N
2781526
C1=CC(=C(C=C1Cl)OC(F)(F)F)N
C7H5ClF3NO
MFCD00153102
4-chloro-2-trifluoromethoxy aniline, 4-chloro-2-trifluoromethoxy phenylamine, benzenamine, 4-chloro-2-trifluoromethoxy, pubchem2960, chlorotrifluoromethoxyphenylamine, benzenamine,4-chloro-2-trifluoromethoxy, 2-amino-5-chlorophenyl trifluoromethyl ether, 2-amino-5-chloro-alpha,alpha,alpha-trifluoroanisole, 4-chloro-2-trifluoromethoxy aniline; 4-chloro-2-trifluoromethoxy phenylamine
4-chloro-2-(trifluoromethoxy)aniline
Safety and Handling

Safety and Handling

TSCA : No

Documents