Please login to your online account to display your discounted pricing

4-Chloro-2-phenylquinazoline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-phenylquinazoline
CAS 6484-25-9
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C14H9ClN2
Formula Weight 240.69g/mol
MDL Number MFCD00006713
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171930250

  SDS
thermo fisher scientific
171930250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171930050

  SDS
fisher scientific acros
171930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Sulfated Ash 0.1% max.
Melting Point 123° to 128°C
Color Slightly yellow to yellow
Quantity 25g
Chemical Name or Material 4-Chloro-2-phenylquinazoline
CAS 6484-25-9
CAS Min % 96.0
CAS Max % 100.0
Physical Form Slightly yellow to yellow crystalline powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C14H9ClN2
Formula Weight 240.69g/mol
MDL Number MFCD00006713
Fieser 04,86
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.