Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-nitrodiphenyl sulfide, 98%

$94.97 - $367.89

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-nitrodiphenyl sulfide
CAS 4548-56-5
Molecular Formula C12H8ClNO2S
Formula Weight 265.72
MDL Number MFCD00052249
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1322506

  SDS
alfa aesar™
A1322506
5g Each for $94.97

AAA1322514

  SDS
alfa aesar™
A1322514
25g Each for $367.89
Description & Specifications

Specifications

Formula Weight 265.72
Chemical Name or Material 4-Chloro-2-nitrodiphenyl sulfide
Assay 98%
Molecular Formula C12H8ClNO2S
MDL Number MFCD00052249
CAS 4548-56-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 82° to 84°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.