Please login to your online account to display your discounted pricing

4-Chloro-2-nitrobenzyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-nitrobenzyl chloride
CAS 938-71-6
Assay Percent Range 98%
Molecular Formula C7H5Cl2NO2
Linear Formula ClC6H3(NO2)CH2Cl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC187950010

  SDS
thermo fisher scientific
AC187950010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC187950050

  SDS
thermo fisher scientific
AC187950050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-Chloro-2-nitrobenzyl chloride
Assay Percent Range 98%
Molecular Formula C7H5Cl2NO2
Linear Formula ClC6H3(NO2)CH2Cl
CAS 938-71-6