missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-2-nitrobenzyl chloride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 187950050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Chloro-2-nitrobenzyl chloride | |
97.5% min. (GC) | |
C7H5Cl2NO2 | |
MFCD00007216 | |
4-chloro-2-nitrobenzyl chloride, 4-chloro-1-chloromethyl-2-nitrobenzene, 4-chloro-2-nitrobenzylchloride, acmc-20amlj, benzene, 4-chloro-1-chloromethyl-2-nitro, 2-nitro-4-chlorobenzyl chloride, omphlgrocarzou-uhfffaoysa, .alpha.,4-dichloro-2-nitrotoluene | |
[O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1 | |
206.02 | |
206.03 |
938-71-6 | |
Glass Bottle | |
ClC6H3(NO2)CH2Cl | |
5g | |
QBIQAXWMONJLNW-UHFFFAOYSA-N | |
1-chloro-3-(chloromethyl)-5-nitrobenzene | |
598399 | |
98% |
Chemical Identifiers
938-71-6 | |
206.02 | |
QBIQAXWMONJLNW-UHFFFAOYSA-N | |
598399 | |
[O-][N+](=O)C1=CC(Cl)=CC(CCl)=C1 |
C7H5Cl2NO2 | |
MFCD00007216 | |
4-chloro-2-nitrobenzyl chloride, 4-chloro-1-chloromethyl-2-nitrobenzene, 4-chloro-2-nitrobenzylchloride, acmc-20amlj, benzene, 4-chloro-1-chloromethyl-2-nitro, 2-nitro-4-chlorobenzyl chloride, omphlgrocarzou-uhfffaoysa, .alpha.,4-dichloro-2-nitrotoluene | |
1-chloro-3-(chloromethyl)-5-nitrobenzene |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 213-345-9
RUO â Research Use Only