missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-2-nitrobenzoic Acid 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 109661000

 View more versions of this product

Catalog No. AC109661000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Powder
201.56
Light yellow to light yellow-green
09, 401
100.0
6280-88-2
ClC6H3(NO2)CO2H
4-Chloro-2-nitrobenzoic acid
MFCD00007214
≥96.0%
100g
141° to 146°C
96.0
Glass bottle
97%
97%
C7 H4 Cl N O4
Authentic

Chemical Identifiers

6280-88-2
100.0
C7 H4 Cl N O4
201.56
4-Chloro-2-nitrobenzoic acid, 97%
96.0
97%
ClC6H3(NO2)CO2H
MFCD00007214
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 228-483-5