Please login to your online account to display your discounted pricing

4-Chloro-2-nitrobenzoic Acid 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-nitrobenzoic acid
Name Note 97%
CAS 6280-88-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7 H4 Cl N O4
Linear Formula ClC6H3(NO2)CO2H
Formula Weight 201.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC109661000

  SDS
thermo fisher scientific
109661000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 201.56
Quantity 100g
Color Light yellow to light yellow-green
Melting Point 141° to 146°C
EINECS Number 228-483-5
Beilstein 09, 401
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 6280-88-2
Assay Percent Range 97%
Linear Formula ClC6H3(NO2)CO2H
Name Note 97%
Chemical Name or Material 4-Chloro-2-nitrobenzoic acid
Molecular Formula C7 H4 Cl N O4
MDL Number MFCD00007214
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.