Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-nitroanisole, 99+%

$38.57 - $551.47

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-nitroanisole
CAS 89-21-4
Molecular Formula C7H6ClNO3
Formula Weight 187.58
MDL Number MFCD00024327
Synonym 4-Chloro-2-nitro-1-methoxybenzene; 4-Methoxy-3-nitrochlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2461006

  SDS
alfa aesar™
B2461006
5g Each for $38.57

AAB2461014

  SDS
alfa aesar™
B2461014
25g Each for $155.20

AAB2461022

  SDS
alfa aesar™
B2461022
100g Each for $551.47
Description & Specifications

Specifications

Formula Weight 187.58
Health Hazard 1 Warning
Assay ≥99%
Synonym 4-Chloro-2-nitro-1-methoxybenzene; 4-Methoxy-3-nitrochlorobenzene
Chemical Name or Material 4-Chloro-2-nitroanisole
CAS 89-21-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 97° to 99°C
Quantity 5g
Molecular Formula C7H6ClNO3
MDL Number MFCD00024327
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.