Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-nitroaniline, 98%

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-nitroaniline
CAS 89-63-4
Molecular Formula C6H5ClN2O2
Formula Weight 172.57
MDL Number MFCD00007836
UN Number UN2237
Beilstein 512436
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA181430B

 
alfa aesar™
A181430B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1814330

 
alfa aesar™
A1814330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA181430I

 
alfa aesar™
A181430I
5000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.57
Chemical Name or Material 4-Chloro-2-nitroaniline
TSCA TSCA
Odor Weak, Characteristic
Assay 98%
UN Number UN2237
Health Hazard 2 GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 191°C (375°F)
Melting Point 114° to 118°C
Molecular Formula C6H5ClN2O2
MDL Number MFCD00007836
CAS 89-63-4
Beilstein 512436
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROANILINES
RTECS Number BX1575000
Quantity 1000g

Applications
4-Chloro-2-nitroaniline is used as pharmaceutical intermediate.

Solubility
Soluble in water (0.5 g/L).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.