missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-nitro-N,N-di-n-propylaniline, 97%

$36.27 - $426.82

Chemical Identifiers

CAS 40832-47-1
Synonym (4-Chloro-2-nitrophenyl)di-n-propylamine
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63709MD
View Documents
Alfa Aesar™
H63709MD
250mg Each for $36.27
 
AAH6370903
View Documents
Alfa Aesar™
H6370903
1g Each for $107.53
 
AAH6370906
View Documents
Alfa Aesar™
H6370906
5g Each for $426.82
 
Specifications

Chemical Identifiers

40832-47-1
(4-Chloro-2-nitrophenyl)di-n-propylamine

Specifications

250mg
UN2811
(4-Chloro-2-nitrophenyl)di-n-propylamine
Solid
4-Chloro-2-nitro-N,N-di-n-propylaniline
40832-47-1
915754
256.73
97%
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures