missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-nitro-N,N-di-n-propylaniline, 97%

Manufacturer:  Alfa Aesar™ H6370906

 View more versions of this product

Catalog No. AAH6370906



Specifications

Specifications

5g
UN2811
(4-Chloro-2-nitrophenyl)di-n-propylamine
Solid
40832-47-1
915754
256.73
97%

Chemical Identifiers

40832-47-1
(4-Chloro-2-nitrophenyl)di-n-propylamine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents