Learn More
4-Chloro-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals
$2263.49 - $2263.49
Chemical Identifiers
CAS | 49844-90-8 |
---|---|
Molecular Formula | C5H5ClN2S |
Molecular Weight (g/mol) | 160.62 |
MDL Number | MFCD00006083 |
InChI Key | DFOHHQRGDOQMKG-UHFFFAOYSA-N |
Synonym | 4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn |
PubChem CID | 97113 |
IUPAC Name | 4-chloro-2-methylsulfanylpyrimidine |
SMILES | CSC1=NC=CC(Cl)=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2148130
|
Thermo Scientific Chemicals
B2148130 |
250 g |
Each for $2,263.49
|
|
|||||
Description
It finds it application as a building block used in medicinal chemistry synthesis. 4-Chloro-2-methylthiopyrimidine is used in the total synthesis of the marine alkaloid variolin B1 and 2-hydroxy-4-pyrimidinecarboxylic acid. It is also used in the synthesis of 2,4-disubstituted pyrimidines, a novel class of KDR kinase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds it application as a building block used in medicinal chemistry synthesis. 4-Chloro-2-methylthiopyrimidine is used in the total synthesis of the marine alkaloid variolin B1 and 2-hydroxy-4-pyrimidinecarboxylic acid. It is also used in the synthesis of 2,4-disubstituted pyrimidines, a novel class of KDR kinase inhibitors.
Solubility
Not miscible or difficult to mix in water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/moisture, heat and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.
Chemical Identifiers
49844-90-8 | |
160.62 | |
DFOHHQRGDOQMKG-UHFFFAOYSA-N | |
97113 | |
CSC1=NC=CC(Cl)=N1 |
C5H5ClN2S | |
MFCD00006083 | |
4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn | |
4-chloro-2-methylsulfanylpyrimidine |
Specifications
49844-90-8 | |
1.38 | |
>110°C (230°F) | |
1.6 00 | |
250 g | |
118042 | |
4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn | |
DFOHHQRGDOQMKG-UHFFFAOYSA-N | |
4-chloro-2-methylsulfanylpyrimidine | |
97113 | |
98% |
∼-2°C | |
118°C to 120°C (9 mmHg) | |
C5H5ClN2S | |
MFCD00006083 | |
UN1760 | |
Moisture sensitive; May form a precipitate | |
Not miscible or difficult to mix in water. | |
CSC1=NC=CC(Cl)=N1 | |
160.62 | |
160.63 | |
4-Chloro-2-(methylthio)pyrimidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 256-500-6
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only