missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine, 97%

Manufacturer:  Alfa Aesar™ H6162006

 View more versions of this product

Catalog No. AAH6162006



Specifications

Specifications

4-Chloro-2-methylthio-6-(trifluoromethyl)pyrimidine
5g
C6H4ClF3N2S
2-methylthio-4-trifluoromethyl-6-chloropyrimidine
CSC1=NC(=CC(=N1)Cl)C(F)(F)F
228.617
228.62
97%
Colorless
16097-63-5
MFCD00269080
GOORQLLDZBAQIW-UHFFFAOYSA-N
4-chloro-2-methylsulfanyl-6-(trifluoromethyl)pyrimidine
248063
Liquid

Chemical Identifiers

16097-63-5
228.617
GOORQLLDZBAQIW-UHFFFAOYSA-N
248063
CSC1=NC(=CC(=N1)Cl)C(F)(F)F
C6H4ClF3N2S
MFCD00269080
2-methylthio-4-trifluoromethyl-6-chloropyrimidine
4-chloro-2-methylsulfanyl-6-(trifluoromethyl)pyrimidine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents