Learn More
4-Chloro-2-methylquinoline, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1500203
Description
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
4-Chloro-2-methylquinoline | |
∼25°C | |
269°C to 270°C | |
C10H8ClN | |
MFCD00006757 | |
3522 | |
Slightly soluble in water. | |
CC1=NC2=CC=CC=C2C(=C1)Cl | |
177.63 | |
177.63 |
4295-06-1 | |
0.881 | |
>110°C (230°F) | |
1.622 | |
1 g | |
4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl | |
HQAIROMRVBVWSK-UHFFFAOYSA-N | |
4-chloro-2-methylquinoline | |
77973 | |
97% |
Chemical Identifiers
4295-06-1 | |
MFCD00006757 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-300-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only