missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylquinoline, 97%

Manufacturer:  Alfa Aesar™ A1500214

 View more versions of this product

Catalog No. AAA1500214



Description

Description

Applications
4-Chloroquinaldine is used in the preparation of 4-phenyl-hydrazinoquinaldine

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

4-Chloro-2-methylquinoline
269°C to 270°C
25g
C10H8ClN
3522
HQAIROMRVBVWSK-UHFFFAOYSA-N
4-chloro-2-methylquinoline
77973
97%
>110°C (230°F)
~25°C
4295-06-1
MFCD00006757
4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
CC1=NC2=CC=CC=C2C(=C1)Cl
177.63
177.63

Chemical Identifiers

4295-06-1
177.63
HQAIROMRVBVWSK-UHFFFAOYSA-N
77973
CC1=NC2=CC=CC=C2C(=C1)Cl
C10H8ClN
MFCD00006757
4-chloroquinaldine, quinoline, 4-chloro-2-methyl, 4-chloro-2-methyl-quinoline, 2-methyl-4-chloroquinoline, quinaldine, 4-chloro, 4-chloro-quinaldine, acmc-20a2ui, 4-chloroquinaldine, 4-chloroquinaldine hcl, quinoline,4-chloro-2-methyl
4-chloro-2-methylquinoline
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents