Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylpyridine hydrochloride, 98%

$102.01 - $1,387.73

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-methylpyridine hydrochloride
CAS 41753-34-8
Molecular Formula C6H6ClN·HCl
Formula Weight 164.04
MDL Number MFCD03092889
Synonym 4-Chloro-2-picoline hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2471903

  SDS
alfa aesar™
B2471903
1g Each for $102.01

AAB2471906

  SDS
alfa aesar™
B2471906
5g Each for $379.70

AAB2471914

  SDS
alfa aesar™
B2471914
25g Each for $1,387.73
Description & Specifications

Specifications

Formula Weight 164.04
Chemical Name or Material 4-Chloro-2-methylpyridine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 41753-34-8
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 207° to 210°C (dec.)
Quantity 1g
Synonym 4-Chloro-2-picoline hydrochloride
MDL Number MFCD03092889
Molecular Formula C6H6ClN·HCl
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.