missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylphenyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1047006

 View more versions of this product

Catalog No. AAL1047006



Specifications

Specifications

4-Chloro-2-methylphenyl isocyanate
103°C (13mmHg)
5g
C8H6ClNO
UN2811
4-chloro-2-methylphenyl isocyanate, 4-chloro-2-methylphenylisocyanate, 4-chloro-o-tolyl isocyanate, benzene, 4-chloro-1-isocyanato-2-methyl, 4-chloro-1-isocyanato-2-methyl-benzene, acmc-20amlp, 4-chloro-2-methylbenzenisocyanate, #, 4-chloranyl-1-isocyanato-2-methyl-benzene
CC1=C(C=CC(=C1)Cl)N=C=O
167.592
167.6
>110°C (230°F)
36°C to 40°C
37408-18-7
MFCD00019914
2803097
FTZJLXIATZSKIL-UHFFFAOYSA-N
4-chloro-1-isocyanato-2-methylbenzene
142157
98%

Chemical Identifiers

37408-18-7
167.592
FTZJLXIATZSKIL-UHFFFAOYSA-N
142157
CC1=C(C=CC(=C1)Cl)N=C=O
C8H6ClNO
MFCD00019914
4-chloro-2-methylphenyl isocyanate, 4-chloro-2-methylphenylisocyanate, 4-chloro-o-tolyl isocyanate, benzene, 4-chloro-1-isocyanato-2-methyl, 4-chloro-1-isocyanato-2-methyl-benzene, acmc-20amlp, 4-chloro-2-methylbenzenisocyanate, #, 4-chloranyl-1-isocyanato-2-methyl-benzene
4-chloro-1-isocyanato-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H330-H334-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents