Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylbenzyl alcohol, 97%

$87.08 - $292.31

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-methylbenzyl alcohol
CAS 129716-11-6
Molecular Formula C8H9ClO
Formula Weight 156.61
MDL Number MFCD06201140
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2755403

  SDS
alfa aesar™
H2755403
1g Each for $87.08

AAH2755406

  SDS
alfa aesar™
H2755406
5g Each for $292.31
Description & Specifications

Specifications

Formula Weight 156.61
Chemical Name or Material 4-Chloro-2-methylbenzyl alcohol
Assay 97%
Molecular Formula C8H9ClO
MDL Number MFCD06201140
CAS 129716-11-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 60° to 64°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.