missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylbenzonitrile, 97%

$16.88 - $183.22

Chemical Identifiers

CAS 50712-68-0
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00045603
InChI Key ZZAJFWXXFZTTLH-UHFFFAOYSA-N
Synonym 2-cyano-5-chlorotoluene
PubChem CID 142742
IUPAC Name 4-chloro-2-methylbenzonitrile
SMILES CC1=C(C=CC(=C1)Cl)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55261MD View Documents Alfa Aesar™
H55261MD
250mg Each for $16.88
 
AAH5526103 View Documents Alfa Aesar™
H5526103
1g Each for $50.62
 
AAH5526106 View Documents Alfa Aesar™
H5526106
5g Each for $183.22
 
Specifications

Chemical Identifiers

50712-68-0
151.593
ZZAJFWXXFZTTLH-UHFFFAOYSA-N
142742
CC1=C(C=CC(=C1)Cl)C#N
C8H6ClN
MFCD00045603
2-cyano-5-chlorotoluene
4-chloro-2-methylbenzonitrile

Specifications

57°C to 61°C
50712-68-0
MFCD00045603
2-cyano-5-chlorotoluene
CC1=C(C=CC(=C1)Cl)C#N
151.593
151.6
4-Chloro-2-methylbenzonitrile
250mg
C8H6ClN
UN3439
ZZAJFWXXFZTTLH-UHFFFAOYSA-N
4-chloro-2-methylbenzonitrile
142742
97%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures