missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylbenzonitrile, 97%

Manufacturer:  Alfa Aesar™ H55261MD

 View more versions of this product

Catalog No. AAH55261MD



Description

Description

Specifications

Specifications

4-Chloro-2-methylbenzonitrile
250mg
C8H6ClN
UN3439
ZZAJFWXXFZTTLH-UHFFFAOYSA-N
4-chloro-2-methylbenzonitrile
142742
97%
57°C to 61°C
50712-68-0
MFCD00045603
2-cyano-5-chlorotoluene, 2-methyl-4-chlorobenzonitrile, 4-chloro-2-methyl benzonitrile, 4-chloro-2-methyl-benzonitrile, 4-chloro-2-methylbenzenecarbonitrile, 5-chloro-2-cyanotoluene, acmc-1awhh, benzonitrile, 4-chloro-2-methyl, ksc494a4r, pubchem12503
CC1=C(C=CC(=C1)Cl)C#N
151.593
151.6

Chemical Identifiers

50712-68-0
151.593
ZZAJFWXXFZTTLH-UHFFFAOYSA-N
142742
CC1=C(C=CC(=C1)Cl)C#N
C8H6ClN
MFCD00045603
2-cyano-5-chlorotoluene, 2-methyl-4-chlorobenzonitrile, 4-chloro-2-methyl benzonitrile, 4-chloro-2-methyl-benzonitrile, 4-chloro-2-methylbenzenecarbonitrile, 5-chloro-2-cyanotoluene, acmc-1awhh, benzonitrile, 4-chloro-2-methyl, ksc494a4r, pubchem12503
4-chloro-2-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents