Learn More
4-chloro-2-methylbenzoic acid, 97%, Maybridge™
Manufacturer: Fisher Scientific Company TL00910EA
Specifications
4-Chloro-2-methylbenzoic acid | |
10g | |
97.0 | |
C8H7ClO2 | |
2-methyl-4-chlorobenzoic acid, 4-chloro-2-methyl-benzoic acid, 4-chloro-2-methylbenzoic acid, 4-chloro-2-methylbenzoicacid, 4-chloro-o-toluic acid, acmc-1bjnb, benzoic acid, 4-chloro-2-methyl, ksc497q4r, o-toluic acid, 4-chloro, pubchem4570 | |
CC1=C(C=CC(=C1)Cl)C(=O)O | |
170.59 | |
170.59 |
Amber Glass Bottle | |
7499-07-2 | |
97% | |
MFCD00045852 | |
XXFKOBGFMUIWDH-UHFFFAOYSA-N | |
4-chloro-2-methylbenzoic acid | |
348269 | |
97% |
Chemical Identifiers
7499-07-2 | |
170.59 | |
XXFKOBGFMUIWDH-UHFFFAOYSA-N | |
348269 | |
CC1=C(C=CC(=C1)Cl)C(=O)O |
C8H7ClO2 | |
MFCD00045852 | |
2-methyl-4-chlorobenzoic acid, 4-chloro-2-methyl-benzoic acid, 4-chloro-2-methylbenzoic acid, 4-chloro-2-methylbenzoicacid, 4-chloro-o-toluic acid, acmc-1bjnb, benzoic acid, 4-chloro-2-methyl, ksc497q4r, o-toluic acid, 4-chloro, pubchem4570 | |
4-chloro-2-methylbenzoic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.