Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methylbenzeneboronic acid, 98%

$664.97 - $2,479.78

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-methylbenzeneboronic acid
CAS 209919-30-2
Molecular Formula C7H8BClO2
Formula Weight 170.4
MDL Number MFCD02683107
Synonym 4-Chloro-2-methylphenylboronic acid; 4-Chloro-o-tolylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2368806

 
alfa aesar™
B2368806
5g Each for $664.97

AAB2368814

 
alfa aesar™
B2368814
25g Each for $2,479.78
Description & Specifications

Specifications

Formula Weight 170.4
Chemical Name or Material 4-Chloro-2-methylbenzeneboronic acid
Assay 98%
Molecular Formula C7H8BClO2
MDL Number MFCD02683107
CAS 209919-30-2
Synonym 4-Chloro-2-methylphenylboronic acid; 4-Chloro-o-tolylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 193° to 195°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.