Please login to your online account to display your discounted pricing

4-Chloro-2-methylaniline, 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC109375000

 View more versions of this product

Catalog No. AC109375000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 141.6
Quantity 500g
Density 1.1900g/mL
CAS Max % 100.0
Synonym 4-Chloro-o-toluidine
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H8ClN
CAS 95-69-2
CAS Min % 97.5
Chemical Name or Material 4-Chloro-2-methylaniline
MDL Number MFCD00007842
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Linear Formula ClC6H3(CH3)NH2
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Toxic if swallowed.
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Chloro-2-methylaniline
Name Note 98%
CAS 95-69-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Formula Weight 141.6
MDL Number MFCD00007842
Synonym 4-Chloro-o-toluidine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
May cause cancer.
Toxic if swallowed.
Suspected of causing genetic defects.

SDS & Safety Documents