Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chloro-2'-methylacetanilide, 97%

$146.77 - $146.77

Chemical Identifiers

Chemical Name or Material 4'-Chloro-2'-methylacetanilide
CAS 5202-86-8
Molecular Formula C9H10ClNO
Formula Weight 183.64
MDL Number MFCD00027399
Beilstein 2087943
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2237109

  SDS
alfa aesar™
B2237109
10g Each for $146.77
Description & Specifications

Specifications

Formula Weight 183.64
Chemical Name or Material 4'-Chloro-2'-methylacetanilide
Assay 97%
Molecular Formula C9H10ClNO
MDL Number MFCD00027399
CAS 5202-86-8
Beilstein 2087943
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 139° to 140°C
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.