Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methyl-7-(trifluoromethyl)quinoline, 97%

$116.99 - $404.82

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-methyl-7-(trifluoromethyl)quinoline
CAS 18529-09-4
Molecular Formula C11H7ClF3N
Formula Weight 245.63
MDL Number MFCD00272334
Synonym 4-Chloro-7-(trifluoromethyl)quinaldine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5055503

 
alfa aesar™
H5055503
1g Each for $404.82

AAH50555MD

 
alfa aesar™
H50555MD
250mg Each for $116.99
Description & Specifications

Specifications

Formula Weight 245.63
Assay 97%
Molecular Formula C11H7ClF3N
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Chloro-2-methyl-7-(trifluoromethyl)quinoline
MDL Number MFCD00272334
CAS 18529-09-4
Synonym 4-Chloro-7-(trifluoromethyl)quinaldine
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.