Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methoxyphenol, 97%

$51.23 - $51.23

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-methoxyphenol
CAS 16766-30-6
Molecular Formula C7H7ClO2
Formula Weight 158.59
MDL Number MFCD00070774
Beilstein 1940516
Synonym 4-Chloroguaiacol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1480606

  SDS
alfa aesar™
A1480606
5g Each for $51.23
Description & Specifications

Specifications

Formula Weight 158.59
Chemical Name or Material 4-Chloro-2-methoxyphenol
Odor Phenol-like
Assay 97%
Molecular Formula C7H7ClO2
MDL Number MFCD00070774
CAS 16766-30-6
Beilstein 1940516
Synonym 4-Chloroguaiacol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point >110°C (230°F)
Boiling Point 241°C
Melting Point 16° to 17°C
Quantity 5g

Applications
4-Chloro-2-methoxyphenol, is used as an intermediate in pharmaceutical research, pharmaceutical chemicals and reagents. It is also used as an application intermediate. it can be used to produce 4-Chloro-2-methoxy-6-nitrophenol.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.