missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-2-methoxyphenol, 97%

Manufacturer:  Alfa Aesar™ A1480614

 View more versions of this product

Catalog No. AAA1480614



Description

Description

Applications
4-Chloro-2-methoxyphenol, is used as an intermediate in pharmaceutical research, pharmaceutical chemicals and reagents. It is also used as an application intermediate. it can be used to produce 4-Chloro-2-methoxy-6-nitrophenol.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases.
Specifications

Specifications

4-Chloro-2-methoxyphenol
Phenol-like
16°C to 17°C
16766-30-6
MFCD00070774
4-chloroguaiacol, phenol, 4-chloro-2-methoxy, 4-chloro-2-methoxy-phenol, 2-methoxy-4-chloro-phenol, pubchem3665, acmc-209dwx, #, phenol,4-chloro-2-methoxy, 4-chloranyl-2-methoxy-phenol, fvzqmmmrfnursh-uhfffaoysa
COC1=C(C=CC(=C1)Cl)O
158.581
158.59
>110°C (230°F)
241°C
25g
C7H7ClO2
1940516
FVZQMMMRFNURSH-UHFFFAOYSA-N
4-chloro-2-methoxyphenol
28050
97%

Chemical Identifiers

16766-30-6
158.581
FVZQMMMRFNURSH-UHFFFAOYSA-N
28050
COC1=C(C=CC(=C1)Cl)O
C7H7ClO2
MFCD00070774
4-chloroguaiacol, phenol, 4-chloro-2-methoxy, 4-chloro-2-methoxy-phenol, 2-methoxy-4-chloro-phenol, pubchem3665, acmc-209dwx, #, phenol,4-chloro-2-methoxy, 4-chloranyl-2-methoxy-phenol, fvzqmmmrfnursh-uhfffaoysa
4-chloro-2-methoxyphenol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

SDS Documents