Please login to your online account to display your discounted pricing

4-Chloro-2-methoxy-5-methylaniline, 85%, Tech., ACROS Organics™


Quantity 50g

Manufacturer: thermo fisher scientific  331360500

 View more versions of this product

Catalog No. AC331360500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥80%
Purity Grade Technical Grade
Physical Form Crystalline Solid
Formula Weight 171.63
Quantity 50g
Color Black
EINECS Number 228-943-5
Melting Point 97° to 103°C
CAS Min % 80.0
CAS Max % 100.0
Chemical Name or Material 4-Chloro-2-methoxy-5-methylaniline
Infrared Spectrum Authentic
Name Note tech., 85%
Assay Percent Range 85%
Molecular Formula C8H10ClNO
MDL Number MFCD00007847
CAS 6376-14-3
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Chloro-2-methoxy-5-methylaniline
Name Note tech., 85%
CAS 6376-14-3
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C8H10ClNO
Formula Weight 171.63
MDL Number MFCD00007847
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

SDS & Safety Documents