Please login to your online account to display your discounted pricing

4-Chloro-2-methoxy-5-methylaniline, 85%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-2-methoxy-5-methylaniline
Name Note tech., 85%
CAS 6376-14-3
CAS Min % 80.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥80%
Assay Percent Range 85%
Molecular Formula C8H10ClNO
Formula Weight 171.63
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC331360050

  SDS
thermo fisher scientific
331360050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥80%
Purity Grade Technical Grade
Formula Weight 171.63
Physical Form Crystalline Solid
Quantity 5g
Color Black
EINECS Number 228-943-5
Melting Point 97° to 103°C
CAS Max % 100.0
CAS Min % 80.0
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Assay Percent Range 85%
Chemical Name or Material 4-Chloro-2-methoxy-5-methylaniline
CAS 6376-14-3
Molecular Formula C8H10ClNO
Name Note tech., 85%
MDL Number MFCD00007847
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.