missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-chloro-2-iodobenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439260010

 View more versions of this product

Catalog No. AC439260010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Chloro-2-iodobenzoic acid
Glass bottle
White
1g
96.0
96% min. (HPLC)
2-iodo-4-chlorobenzoic acid, 4-chloro-2-iodo-benzoic acid, 4-chloro-2-iodobenzoic acid, 4-chloro-2-iodobenzoicacid, acmc-209bu6, benzoic acid, 4-chloro-2-iodo, benzoic acid,4-chloro-2-iodo, buttpark 100\07-49, ksc519a3p, pubchem4018
C1=CC(=C(C=C1Cl)I)C(=O)O
282.47
282.47
97%
Authentic
3% max.
165.2°C to 166.4°C
13421-13-1
100.0
C7H4ClIO2
LRRDANNSUCQNDU-UHFFFAOYSA-N
4-chloro-2-iodobenzoic acid
139452
Crystalline Powder

Chemical Identifiers

13421-13-1
282.47
2-iodo-4-chlorobenzoic acid, 4-chloro-2-iodo-benzoic acid, 4-chloro-2-iodobenzoic acid, 4-chloro-2-iodobenzoicacid, acmc-209bu6, benzoic acid, 4-chloro-2-iodo, benzoic acid,4-chloro-2-iodo, buttpark 100\07-49, ksc519a3p, pubchem4018
4-chloro-2-iodobenzoic acid
C7H4ClIO2
LRRDANNSUCQNDU-UHFFFAOYSA-N
139452
C1=CC(=C(C=C1Cl)I)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS Documents